2-(Benzyloxy)-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

131703-500MG 131703-1G 131703-5G
77337-640EA 432.6 USD
77337-640 77347-890 77372-014
2-(Benzyloxy)-3-formylbenzoic acid ≥97%
2-(Benzyloxy)-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₄
MW: 256.2533 g/mol
Storage Temperature: Ambient
CAS Number: 909704-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR