3-Bromo-5-chloro-2-(methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

131579-500MG 131579-1G 131579-25G 131579-5G
77323-872EA 285.31 USD
77323-872 77347-888 77383-758 77372-012
3-Bromo-5-chloro-2-(methylthio)thiophene ≥97%
3-Bromo-5-chloro-2-(methylthio)thiophene

Precaución: for Research Use Only

Formula: C₅H₄BrClS₂
MW: 243.5722 g/mol
Storage Temperature: Ambient
CAS Number: 2918954-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR