2-Amino-4-chloro-3-fluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
131133-500MG
131133-1G
131133-5G
131133-25G
77323-870EA
221.45
USD
77323-870
77347-886
77360-056
77383-756
2-Amino-4-chloro-3-fluorobenzoic acid ≥97%
2-Amino-4-chloro-3-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅ClFNO₂ MW: 189.5715 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11193652 CAS Number: 1039878-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...