2,5-Dimethylthiophene-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

131114-500MG 131114-1G 131114-5G 131114-25G
77323-868EA 83.43 USD
77323-868 77342-184 77360-054 77372-010
2,5-Dimethylthiophene-3-carboxylic acid ≥97%
2,5-Dimethylthiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860238
CAS Number: 26421-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR