2'-Bromo-2-methoxy-4-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

130337-500MG 130337-1G 130337-5G 130337-25G
77323-866EA 251.32 USD
77323-866 77347-884 77360-052 77383-754
2'-Bromo-2-methoxy-4-methyl-1,1'-biphenyl ≥97%
2'-Bromo-2-methoxy-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.1564 g/mol
Storage Temperature: Ambient
CAS Number: 2918957-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR