2-Chloro-6-fluoro-4-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

130299-500MG 130299-1G 130299-5G
77337-244EA 408.91 USD
77337-244 77347-882 77372-008
2-Chloro-6-fluoro-4-methoxyphenol ≥95%
2-Chloro-6-fluoro-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO₂
Storage Temperature: Ambient
CAS Number: 1600530-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR