2-Fluoro-6-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

130056-1G 130056-5G 130056-25G
77341-540EA 91.67 USD
77341-540 77360-050 77372-006
2-Fluoro-6-(trifluoromethoxy)benzoic acid ≥97%
2-Fluoro-6-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
MW: 224.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22551466
CAS Number: 1211530-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR