8-(2,4-Dichlorophenyl)-1,4-Dioxaspiro[4.5]decan-8-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
130004-500MG
130004-1G
130004-5G
130004-25G
77323-862EA
202.91
USD
77323-862
77347-880
77360-046
77383-750
8-(2,4-Dichlorophenyl)-1,4-Dioxaspiro[4.5]decan-8-ol ≥97%
8-(2,4-Dichlorophenyl)-1,4-Dioxaspiro[4.5]decan-8-ol
Precaución: for Research Use Only
Formula:
C₁₄H₁₆Cl₂O₃ Storage Temperature: Ambient |
CAS Number:
1880070-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...