8-(2-ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

129962-500MG 129962-1G 129962-5G 129962-25G
77323-860EA 161.71 USD
77323-860 77346-066 77360-044 77383-748
8-(2-ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2-ethylphenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₃
MW: 262.344 g/mol
Storage Temperature: Ambient
CAS Number: 1819366-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR