1,2-Dimethyl-3-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-1-iodo-3,4-dimethylbenzene

129846-500MG 129846-1G 129846-5G 129846-25G
77323-858EA 99.91 USD
77323-858 77342-910 77360-042 77372-004
1,2-Dimethyl-3-fluoro-4-iodobenzene ≥97%
1,2-Dimethyl-3-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈FI
MW: 250.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794424
CAS Number: 1806323-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR