5-Bromoisoquinoline 2-oxide ≥97%

Supplier: AOBChem USA

Warning

129801-500MG 129801-1G 129801-5G 129801-25G
77323-856EA 89.61 USD
77323-856 77342-376 77360-038 77372-000
5-Bromoisoquinoline 2-oxide ≥97%
5-Bromoisoquinoline 2-oxide

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Storage Temperature: Ambient
CAS Number: 223671-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR