7-Bromo-6-chloro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

129421-500MG 129421-1G 129421-5G
77323-850EA 312.09 USD
77323-850 77347-876 77371-996
7-Bromo-6-chloro-1H-indazole ≥95%
7-Bromo-6-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
Storage Temperature: Ambient
CAS Number: 1427361-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR