2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

12935-500MG 12935-1G 12935-5G 12935-25G
77323-848EA 230.72 USD
77323-848 77347-874 77360-032 77383-744
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-(2,5-Dimethyl-1H-pyrrol-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₇H₂₃BN₂O₂
MW: 298.188 g/mol
Storage Temperature: Ambient
CAS Number: 885226-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR