2-(Cyclopropylmethoxy)-4-ethyl-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

129257-500MG 129257-1G 129257-5G 129257-25G
77323-846EA 176.13 USD
77323-846 77346-570 77360-030 77383-742
2-(Cyclopropylmethoxy)-4-ethyl-1-methoxybenzene ≥95%
2-(Cyclopropylmethoxy)-4-ethyl-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
CAS Number: 1369845-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR