3-Iodophenetole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Ethoxy-3-iodobenzene

128881-500MG 128881-1G 128881-5G 128881-25G
77323-842EA 121.54 USD
77323-842 77343-972 77360-026 77371-994
3-Iodophenetole ≥97%
3-Iodophenetole

Precaución: for Research Use Only

Formula: C₈H₉IO
MW: 248.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00673013
CAS Number: 29052-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR