3-Bromo-5-chloro-2-fluoro-6-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

128682-500MG 128682-1G 128682-5G
77338-738EA 511.91 USD
77338-738 77347-872 77371-990
3-Bromo-5-chloro-2-fluoro-6-hydroxybenzoic acid ≥97%
3-Bromo-5-chloro-2-fluoro-6-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₃
Storage Temperature: Ambient
CAS Number: 1864362-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR