2-hydroxy-5-iodo-3-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

128661-1G 128661-5G 128661-25G
77341-380EA 82.4 USD
77341-380 77360-022 77371-988
2-hydroxy-5-iodo-3-methylbenzoic acid ≥95%
2-hydroxy-5-iodo-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₃
MW: 278.044 g/mol
Storage Temperature: Ambient
CAS Number: 6174-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR