(2-Bromo-6-fluorophenyl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

128338-500MG 128338-1G 128338-5G 128338-25G
77323-840EA 161.71 USD
77323-840 77346-064 77360-018 77383-738
(2-Bromo-6-fluorophenyl)(furan-2-yl)methanol ≥97%
(2-Bromo-6-fluorophenyl)(furan-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₈BrFO₂
Storage Temperature: Ambient
CAS Number: 1600188-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR