(3-Bromo-2,6-difluorophenyl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

128335-500MG 128335-1G 128335-5G 128335-25G
77323-838EA 182.31 USD
77323-838 77347-104 77360-016 77383-736
(3-Bromo-2,6-difluorophenyl)(furan-2-yl)methanol ≥97%
(3-Bromo-2,6-difluorophenyl)(furan-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₇BrF₂O₂
Storage Temperature: Ambient
CAS Number: 1694821-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR