(5-Bromo-2-fluorophenyl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

128333-500MG 128333-1G 128333-5G 128333-25G
77323-836EA 182.31 USD
77323-836 77347-102 77360-014 77383-734
(5-Bromo-2-fluorophenyl)(furan-2-yl)methanol ≥97%
(5-Bromo-2-fluorophenyl)(furan-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₈BrFO₂
Storage Temperature: Ambient
CAS Number: 1339687-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR