(3-Bromo-2-fluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

128331-500MG 128331-1G 128331-5G 128331-25G
77323-834EA 182.31 USD
77323-834 77347-100 77360-012 77383-732
(3-Bromo-2-fluorophenyl)(furan-2-yl)methanol ≥95%
(3-Bromo-2-fluorophenyl)(furan-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₈BrFO₂
MW: 271.082 g/mol
Storage Temperature: Ambient
CAS Number: 1695268-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR