(5-Chloro-2-fluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

128327-500MG 128327-1G 128327-5G 128327-25G
77323-832EA 182.31 USD
77323-832 77347-098 77360-010 77383-730
(5-Chloro-2-fluorophenyl)(furan-2-yl)methanol ≥95%
(5-Chloro-2-fluorophenyl)(furan-2-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₈ClFO₂
Storage Temperature: Ambient
CAS Number: 1597168-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR