1-(5-Chloro-2-fluorophenyl)cycloheptanol ≥95%

Supplier: AOBChem USA

Warning

128289-500MG 128289-1G 128289-5G
77323-828EA 345.05 USD
77323-828 77347-868 77371-984
1-(5-Chloro-2-fluorophenyl)cycloheptanol ≥95%
1-(5-Chloro-2-fluorophenyl)cycloheptanol

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClFO
MW: 242.717 g/mol
Storage Temperature: Ambient
CAS Number: 2918965-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR