4-Cyano-2,5-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

127640-500MG 127640-1G 127640-5G
77340-306EA 899.19 USD
77340-306 77347-866 77371-978
4-Cyano-2,5-difluorobenzoic acid ≥97%
4-Cyano-2,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₃F₂NO₂
Storage Temperature: Ambient
CAS Number: 198280-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR