(6-Bromo-2,3-dimethoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

127473-500MG 127473-1G 127473-5G 127473-25G
77323-826EA 204.97 USD
77323-826 77347-864 77360-002 77383-726
(6-Bromo-2,3-dimethoxyphenyl)methanol ≥95%
(6-Bromo-2,3-dimethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃
MW: 247.086 g/mol
Storage Temperature: Ambient
CAS Number: 107290-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR