1-Bromo-4-(4-chlorophenoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

127385-500MG 127385-1G 127385-5G 127385-25G
77323-824EA 87.55 USD
77323-824 77342-320 77360-000 77371-976
1-Bromo-4-(4-chlorophenoxy)benzene ≥97%
1-Bromo-4-(4-chlorophenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₂H₈BrClO
Storage Temperature: Ambient
CAS Number: 30427-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR