5-Cyclopentyl-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

127272-500MG 127272-1G 127272-5G
77339-722EA 618 USD
77339-722 77347-858 77371-970
5-Cyclopentyl-2-fluorobenzoic acid ≥97%
5-Cyclopentyl-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃FO₂
Storage Temperature: Ambient
CAS Number: 1539774-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR