(5-Cyclopentyl-2-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

127269-500MG 127269-1G 127269-5G
77339-644EA 605.64 USD
77339-644 77347-856 77371-968
(5-Cyclopentyl-2-fluorophenyl)boronic acid ≥97%
(5-Cyclopentyl-2-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄BFO₂
MW: 208.037 g/mol
Storage Temperature: Ambient
CAS Number: 2225170-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR