4-Bromo-5-fluoro-2-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

127133-500MG 127133-1G 127133-5G
77323-822EA 348.14 USD
77323-822 77347-852 77371-964
4-Bromo-5-fluoro-2-methoxybenzonitrile ≥97%
4-Bromo-5-fluoro-2-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD23707399
CAS Number: 1427360-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR