1-(1-Fluoronaphthalen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

127032-500MG 127032-1G 127032-5G 127032-25G
77323-820EA 182.31 USD
77323-820 77347-096 77359-998 77383-724
1-(1-Fluoronaphthalen-2-yl)ethanone ≥97%
1-(1-Fluoronaphthalen-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₉FO
MW: 188.198 g/mol
Storage Temperature: Ambient
CAS Number: 33718-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR