1-Fluoronaphthalene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Fluoro-2-naphthoic acid

127024-500MG 127024-1G 127024-25G 127024-5G
77323-818EA 291.49 USD
77323-818 77347-850 77383-722 77371-962
1-Fluoronaphthalene-2-carboxylic acid ≥97%
1-Fluoronaphthalene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₇FO₂
MW: 190.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16990867
CAS Number: 574-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR