1-Chloro-3-(4-nitrophenoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

127017-1G 127017-5G 127017-25G
77341-612EA 95.79 USD
77341-612 77359-994 77371-958
1-Chloro-3-(4-nitrophenoxy)benzene ≥97%
1-Chloro-3-(4-nitrophenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₂H₈ClNO₃
MW: 249.65 g/mol
Storage Temperature: Ambient
CAS Number: 2303-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR