1-Bromo-5-fluoro-4-methoxy-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

126963-500MG 126963-1G 126963-5G
77323-816EA 319.3 USD
77323-816 77347-846 77371-952
1-Bromo-5-fluoro-4-methoxy-2-(methoxymethyl)benzene ≥97%
1-Bromo-5-fluoro-4-methoxy-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
CAS Number: 2921705-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR