(2-Bromo-4-fluoro-5-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

126962-500MG 126962-1G 126962-25G 126962-5G
77323-814EA 299.73 USD
77323-814 77347-844 77383-720 77371-950
(2-Bromo-4-fluoro-5-methoxyphenyl)methanol ≥95%
(2-Bromo-4-fluoro-5-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
CAS Number: 1421503-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR