4,5-Dibromo-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

126913-500MG 126913-1G 126913-5G 126913-25G
77323-812EA 121.54 USD
77323-812 77343-896 77359-992 77371-948
4,5-Dibromo-2-methoxybenzoic acid ≥97%
4,5-Dibromo-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₃
MW: 309.939 g/mol
Storage Temperature: Ambient
CAS Number: 1239783-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR