3-Bromo-5-iodobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-5-iodophenyl)methanol

126780-500MG 126780-1G 126780-5G 126780-25G
77323-808EA 150.38 USD
77323-808 77345-378 77359-984 77383-716
3-Bromo-5-iodobenzyl alcohol ≥97%
3-Bromo-5-iodobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09745849
CAS Number: 188813-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR