1-Bromo-4-ethynyl-2,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

126584-500MG 126584-1G
77340-444EA 1079.44 USD
77340-444 77347-842
1-Bromo-4-ethynyl-2,3-difluorobenzene ≥97%
1-Bromo-4-ethynyl-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₈H₃BrF₂
MW: 217.0102 g/mol
Storage Temperature: Ambient
CAS Number: 2000002-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR