2-(Cyclopent-1-en-1-yl)-5-(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

126565-500MG 126565-1G 126565-5G 126565-25G
77323-804EA 145.23 USD
77323-804 77345-118 77359-980 77383-712
2-(Cyclopent-1-en-1-yl)-5-(methylthio)thiophene ≥95%
2-(Cyclopent-1-en-1-yl)-5-(methylthio)thiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₂S₂
MW: 196.3322 g/mol
Storage Temperature: Ambient
CAS Number: 2918954-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR