(E)-3-bromo-5-chlorobenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

126188-500MG 126188-1G 126188-5G
77323-802EA 327.54 USD
77323-802 77347-838 77371-938
(E)-3-bromo-5-chlorobenzaldehyde oxime ≥97%
(E)-3-bromo-5-chlorobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₅BrClNO
MW: 234.478 g/mol
Storage Temperature: Ambient
CAS Number: 1379426-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR