3-Chloro-2-hydroxy-5-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

126124-500MG 126124-1G 126124-5G 126124-25G
77323-800EA 206 USD
77323-800 77347-836 77359-976 77383-710
3-Chloro-2-hydroxy-5-iodobenzaldehyde ≥97%
3-Chloro-2-hydroxy-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClIO₂
Storage Temperature: Ambient
CAS Number: 775333-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR