6-Bromo-4-methoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

126067-500MG 126067-1G 126067-5G 126067-25G
77323-798EA 257.5 USD
77323-798 77347-834 77359-974 77383-708
6-Bromo-4-methoxynicotinic acid ≥97%
6-Bromo-4-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD13189166
CAS Number: 1060805-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR