2-Bromo-4,6-dichloro-3-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

126028-500MG 126028-1G 126028-5G 126028-25G
77323-796EA 232.78 USD
77323-796 77347-832 77359-972 77383-706
2-Bromo-4,6-dichloro-3-(trifluoromethyl)aniline ≥97%
2-Bromo-4,6-dichloro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂F₃N
MW: 308.911 g/mol
Storage Temperature: Ambient
CAS Number: 2918950-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR