3-Bromo-2-iodo-5-nitroanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-iodo-3-methoxy-5-nitrobenzene

125990-500MG 125990-1G 125990-5G 125990-25G
77323-794EA 192.61 USD
77323-794 77347-830 77359-970 77383-704
3-Bromo-2-iodo-5-nitroanisole ≥97%
3-Bromo-2-iodo-5-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrINO₃
Storage Temperature: Ambient
MDL Number: MFCD11846079
CAS Number: 1160574-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR