2,3-Dibromo-5-nitroanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1,2-Dibromo-3-methoxy-5-nitrobenzene

125988-500MG 125988-1G 125988-5G 125988-25G
77323-792EA 134.93 USD
77323-792 77344-506 77359-968 77383-702
2,3-Dibromo-5-nitroanisole ≥95%
2,3-Dibromo-5-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₃
MW: 310.928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846080
CAS Number: 95970-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR