2-(Benzyloxy)-1-chloro-3-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

125909-500MG 125909-1G 125909-5G 125909-25G
77323-790EA 93.73 USD
77323-790 77342-498 77359-966 77371-934
2-(Benzyloxy)-1-chloro-3-fluoro-4-iodobenzene ≥97%
2-(Benzyloxy)-1-chloro-3-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉ClFIO
Storage Temperature: Ambient
CAS Number: 2918951-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR