2-Methyl-1-(thiophen-2-yl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

125572-500MG 125572-1G 125572-5G 125572-25G
77323-786EA 121.54 USD
77323-786 77343-894 77359-962 77371-930
2-Methyl-1-(thiophen-2-yl)propan-1-ol ≥97%
2-Methyl-1-(thiophen-2-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₈H₁₂OS
MW: 156.245 g/mol
Storage Temperature: Ambient
CAS Number: 74641-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR