8-(3-Bromo-2-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

125467-500MG 125467-1G 125467-5G 125467-25G
77323-784EA 223.51 USD
77323-784 77347-828 77359-960 77383-700
8-(3-Bromo-2-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(3-Bromo-2-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₆BrFO₃
Storage Temperature: Ambient
CAS Number: 1881857-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR