8-(5-Bromo-2-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
125460-500MG
125460-1G
125460-5G
125460-25G
77323-782EA
223.51
USD
77323-782
77347-826
77359-958
77383-698
8-(5-Bromo-2-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(5-Bromo-2-fluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol
Precaución: for Research Use Only
Formula:
C₁₄H₁₆BrFO₃ Storage Temperature: Ambient |
CAS Number:
1514868-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...