3-Fluoro-4-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

125261-500MG 125261-1G 125261-5G
77338-864EA 526.33 USD
77338-864 77347-822 77371-926
3-Fluoro-4-iodo-2-methoxypyridine ≥97%
3-Fluoro-4-iodo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅FINO
MW: 253.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611140
CAS Number: 1227595-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR