3-Bromo-2,4-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

125244-500MG 125244-1G 125244-5G 125244-25G
77323-780EA 195.7 USD
77323-780 77347-820 77359-956 77383-696
3-Bromo-2,4-dimethylbenzoic acid ≥95%
3-Bromo-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
CAS Number: 1255206-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR